Safety cans for liquid disposal
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
HDPE (red or white), with stainless steel hardware.
- Chemical resistance and leak protection
- Broad-base design provides stability and resists accidental tip over
- Fixed position on handle allows lid to stay open for easy pouring
Liquid disposal safety cans are the safe, complaint way to dispose liquid flammable or corrosive. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig. Safety can is equipped with a chemically resistant stainless steel flame arrester guards against outside ignition. Carbon insert embedded into the rib of containers provides proper grounding and prevents the creation of a static arc during filling or pouring.