Order Entry
Germany
ContactUsLinkComponent
 
Ammonium nitrate solution 1 mol/l acc. to DIN 19730, Supelco®
  1.02596.2500
 :  Merck
 :  
Ammonium nitrate solution 1 mol/l acc. to DIN 19730, Supelco®
  1.02596.2500
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    100 °C (1013 hPa)
  • Density:
    1.03 g/cm3 (20 °C)
  • Storage temeprature:
    +15...+25 °C
  • Assay:
    0,99 - 1,01 mol/l
  • pH-value:
    5 - 6
  • As (Arsenic):
    ≤0,005 ppm
  • Be (Beryllium):
    ≤0,001 ppm
  • Cd (Cadmium):
    ≤0,002 ppm
  • Co (Cobalt):
    ≤0,005 ppm
  • Cr (Chromium):
    ≤0,005 ppm
  • Cu (Copper):
    ≤0,005 ppm
  • Hg (Mercury):
    ≤0,001 ppm
  • Mg (Magnesium):
    ≤0,001 ppm
  • Mo (Molybdenum):
    ≤0,005 ppm
  • Ni (Nickel):
    ≤0,010 ppm
  • Pb (Lead):
    ≤0,005 ppm
  • Sb (Antimony):
    ≤0,005 ppm
  • Tl (Thallium):
    ≤0,005 ppm
  • U (Uranium):
    ≤0,001 ppm
  • V (Vanadium):
    ≤0,005 ppm
  • Zn (Zinc):
    ≤0,005 ppm

 

 

 :
 :80.04 g/mol
 :M-Clarity™ quality level = MQ100
 :6484-52-2
 :NH₄NO₃
 :MFCD00011425