Cyanoacetylurea ≥98.0% (by HPLC) for synthesis
8.43302.0025
: Merck
:
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Assay (HPLC, area%):≥ 98.0 % (a/a)
- Melting range (lower value):≥ 203 °C
- Melting range (upper value):≤ 207 °C
- Identity (IR):passes test
- HS Code:2926 90 95
- Melting Point:203...207 °C
- Storage:15...25 °C
: N-Carbamoyl-2-cyanoacetamide, Acetamide, N-(aminocarbonyl)-2-cyano-
:
:127.10 g/mol
:6.1,III
:M-Clarity™ quality level = MQ200
:215-906-3
:3439
:GHS06
:Ambient
:1448-98-2
:C₄H₅N₃O₂
:MFCD00007947