Order Entry
Germany
ContactUsLinkComponent
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine for synthesis, Sigma-Aldrich®
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine for synthesis, Sigma-Aldrich®
Catalog # 8.18484.0100
Supplier:  Merck
CAS Number:  
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine for synthesis, Sigma-Aldrich®
Catalog # 8.18484.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Hill Formula:
    C₁₁H₂₁NO₅S
  • Molar Mass:
    279.36 g/mol
  • Flash point:
    110 °C
  • Melting Point:
    147 - 149 °C
  • pH value:
    6.3 (100 g/l, H₂O, 20 °C)
  • Bulk density:
    400 kg/m³
  • Solubility:
    500 g/l (20 °C)
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 3 highly water endangering

Specification Test Results

About this item

 :
 :279.351 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:222-860-8
CAS Number:3637-26-1
Formula:C₁₁H₂₁NO₅S
MDL Number:MFCD00040574