Order Entry
Germany
ContactUsLinkComponent
Cholesteryl chloride for synthesis, Sigma-Aldrich®
Cholesteryl chloride for synthesis, Sigma-Aldrich®
Catalog # 8.41733.0010
Supplier:  Merck
CAS Number:  
Cholesteryl chloride for synthesis, Sigma-Aldrich®
Catalog # 8.41733.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    542 g/l (20 °C)
  • Melting Point:
    712 °C
  • Molar Mass:
    95.22 g/mol
  • Boiling Point:
    1412 °C (1013 hPa)
  • Density:
    2.32 g/cm3 (20 °C)
  • pH:
    =7 (50 g/l, H2O, 20 °C)
  • Synonyms:
    Magnesium dichloride
  • Hill Formula:
    Cl2Mg
  • Chemical Formula:
    MgCl2
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :405.107 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:213-004-4
CAS Number:910-31-6
Formula:C₂₇H₄₅Cl
MDL Number:MFCD00003632