Order Entry
Germany
ContactUsLinkComponent
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride for synthesis, Sigma-Aldrich®
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.24456.0500
Supplier:  Merck
CAS Number:  
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.24456.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    143 - 146 °C
  • Molar Mass:
    195.64 g/mol
  • Synonyms:
    L-4-Hydroxyproline ethyl ester hydrochloride
  • Hill Formula:
    C7H14ClNO3

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :195.643 g/mol
Storage Temperature:Ambient
EINECS:251-779-0
CAS Number:33996-30-4
 :MFCD11506578
Formula:C₇H₁₃NO₃·HCl
MDL Number:MFCD00040589