Order Entry
Germany
ContactUsLinkComponent
 
4,4'-Methylenebis(2,6-di-tert-butylphenol) for synthesis, Sigma-Aldrich®
 :  Merck
 
4,4'-Methylenebis(2,6-di-tert-butylphenol) for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
     - 0.0001 g/l (20 °C)
  • Melting Point:
    153 - 156 °C
  • Molar Mass:
    424.67 g/mol
  • Boiling Point:
    217 °C (1 hPa)
  • Vapor Pressure:
     - 0.1 hPa (25 °C)
  • Flash Point:
    222 °C
  • Density:
    0.99 g/cm3 (20 °C)
  • Storage temperature:
    15...25 °C
  • Assay (GC, area%):
    ≥98% (a/a)
  • Melting range (lower value):
    ≥153 °C
  • Melting range (upper value):
    ≤156 °C
  • Identity (IR):
    passes test