Order Entry
Germany
ContactUsLinkComponent
N-tert-Butyl-ɑ-phenylnitrone for synthesis, Sigma-Aldrich®
N-tert-Butyl-ɑ-phenylnitrone for synthesis, Sigma-Aldrich®
Catalog # 8.41179.0001
Supplier:  Merck
CAS Number:  
N-tert-Butyl-ɑ-phenylnitrone for synthesis, Sigma-Aldrich®
Catalog # 8.41179.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    71 - 73 °C
  • Molar Mass:
    177.24 g/mol
  • HS Code:
    2929 90 00
  • Storage temperature:
    Store below +15 °C
  • Assay (HPLC, area%):
    ≥98% (a/a)
  • Melting range (lower value):
    ≥71 °C
  • Melting range (upper value):
    ≤73 °C
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :177.25 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:222-168-6
CAS Number:3376-24-7
Formula:C₁₁H₁₅NO
MDL Number:MFCD00008799