4-Piperidone monohydrate hydrochloride for synthesis, Sigma-Aldrich®
: Merck
:320589-77-3
:40064-34-4
:C₅H₉NO·xHCl·yH₂O
:MFCD00012776
:M-Clarity™ quality level = MQ200
:99.13 g/mol (free base)
:Ambient
:255-602-8
- Melting Point:95 - 99 °C
- Molar Mass:153.61 g/mol
- Hill Formula:C5H9NO*ClH*H2O
- HS Code:2933 39 99
- Storage temperature:15...25 °C
- Assay (argentometric):≥98% (m)
- Melting range (lower value):≥91 °C
- Melting range (upper value):≤94 °C
- Identity (IR):conforms