Order Entry
Germany
ContactUsLinkComponent
Petroleum spirit, 35…60 °C, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Health
Irritant
Environment
Petroleum spirit, 35…60 °C, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9268-09
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Petroleum spirit, 35…60 °C, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9268-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    275 lb
  • Seq:
    7221

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 10
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.001%
  • ACS - Acidity:
    Passes Test
  • ACS - Boiling Range(Lower):
    35 - 60 °C
  • ACS - Boiling Range(Upper):
    35 - 60 °C

Specification Test Results

About this item

 :
Boiling Pt:35 - 60 °C (1013 hPa)
Density:~0.68 kg (20 °C)
Flash Pt:−18 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :101316-46-5
MDL Number:MFCD00081849