N-Ethyl-p-toluenesulphonamide for synthesis
: Merck
:80-39-7
:
:C₉H₁₃NO₂S
:MFCD00048511
:199.27 g/mol
:M-Clarity™ quality level = MQ200
:Ambient
:201-275-1
- Hill Formula:C₉H₁₃NO₂S
- Molar Mass:199.27 g/mol
- Density:1.26 g/cm³
- Flash point:127 °C
- Melting Point:63 - 65 °C
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 3 highly water endangering
- Storage temperature:15...25 °C