Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. ∼ 70000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol, M.W. ∼ 70000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyvinyl alcohol, M.W. ∼ 70000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    200 °C
  • Bulk Density:
    400 - 600 kg/m3
  • Density:
    1.3 g/cm3 (20 °C)
  • pH:
    4.5 - 7.0 (40 g/l, H2O, 23 °C)
  • Ignition Point:
    230 °C
  • Storage temperature:
    +15...+25 °C
  • Degree of hydrolysis:
    ≥85%
  • dyn.Viscosity:
    6,8 - 9,2 mPa∙s
  • Loss on drying (110 °C, 2 h):
    ≤5,0%
  • Ash:
    ≤1,0%
  • Ester value:
    130 - 150

Specification Test Results

Related Information