Polyvinyl alcohol, M.W. ∼ 30000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
:M-Clarity™ quality level = MQ200
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:200 °C
- Bulk Density:400 - 600 kg/m3
- Density:1.3 g/cm3 (20 °C)
- pH:4.5 - 7.0 (40 g/l, H2O, 23 °C)
- Ignition Point:230 °C
- Storage temperature:15...25 °C
- Degree of hydrolysis:≥85%
- dyn.Viscosity:3,4 - 4,6 mPa∙s
- Loss on drying (110 °C, 2 h):≤5,0%
- Ash:≤ 1,0%
- Ester value:130 - 150