ContactUsLinkComponent
Retention time standards
Retention time standards
Catalog # REST31200
Supplier:  Restek
CAS Number:  
Retention time standards
Catalog # REST31200
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Leaking underground storage tank retention time standard includes n-hexane (C6), n-octacosane (C28), n-decane (C10), n-triacontane (C30), n-dodecane (C12), n-tetracontane (C40), n-tetracosane (C24), with 25 µg/ml each in methylene chloride, 1 ml/ampule
Specifications
About this item
Used during initial sample screening to determine retention time windows for each petroleum product.