Bench cans for solvents
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
These bench cans for solvent safety have plated steel dasher which work as an effective flame arrester to reduce the risk of fire flashback. Hinged cover minimises evaporation losses and protected rim reduces the possibility of hand injury.
- Sturdy 24-gauge steel body construction with tough powder-coat finish
- Compliance with FM and TUV approved