Plunger dispensing cans
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
These plunger cans are used to dispense flammable liquids and solvents safely, without waste or exposure. The pump base is designed to leave less liquid in the bottom before having to refill. Perforated upper pan serves as a flame arrester and top rim has molded edge protector for smooth, trouble-free moistening.
- Chemically resistant Ryton® and brass plunger
- Galvanised steel body construction
Compliance with FM and TÜV approved.
Limited Time Sale
Select items below are currently on sale