ContactUsLinkComponent
Blades for safety knives, No. 60099
Blades for safety knives, No. 60099
  223-0112
undefined
Blades for safety knives, No. 60099
  223-0112
  • Description:
    Spare trapezoid blade (no. 60099), two-sided deep edge, rounded tips
  • L×W:
    55,5×19 mm
  • Thickness:
    0,63 mm
  • Pk:
    1 SET
Trapezoid blade with rounded tips for use with safety knives.
  • Deep-edged, 2-sided ground-edge
  • Double bevel edge-ground
  • 2-times usable blade
Mountable in:
SECUPRO MAXISAFE
SECUPRO MEGASAFE
SECUPRO 625
SECUNORM MULTISET
SECUNORM PROFI25