Specifications
- Antibody type:Primary
- Antigen name:Myosin 1A
- Antigen symbol:MYO1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:4640
- Antigen synonyms:MIHC|DFNA48|BBMI|myosin I heavy chain|MYHL|myosin-Ia|heavy polypeptide-like (100kD)|myosin|brush border myosin I
- Storage buffer:0.1M tris glycine, 20% glycerol, 0.01% thimerosal, pH 7
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:0.68 mg/ml
- Immunogen:Recombinant fragment corresponding to a region within amino acids 299 and 547 of Human myosin IA
- Tested applications:Immunohistochemistry (Paraffin)
- Purification:Antigen affinity chromatography
- Size:100 µL
- Pk:100 µl
Specifications
About this item
This Antibody targets myosin IA in IHC (P) and WB applications and shows reactivity with Human samples. The This Antibody targets each myosin to its particular subcellular location. The kidney epithelial cell line, LLC-PK1-CL4 (CL4), forms a well ordered brush border (BB) on its apical surface. Experiments indicate that the brush border population of the encoded protein turns over rapidly, while its head and tail domains interact transiently with the core actin and plasma membrane, respectively. A rapidly exchanging pool of the protein encoded by this gene envelops an actin core bundle that, by comparison, is static in structure.
Western Blot: 1:500-1:3000, Immunohistochemistry (Paraffin): 1:100-1:1000
Type: Primary
Antigen: myosin IA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human