Order Entry
Germany
ContactUsLinkComponent
Sodium sulphanilate dihydrate sodium salt for synthesis, Sigma-Aldrich®
Sodium sulphanilate dihydrate sodium salt for synthesis, Sigma-Aldrich®
Catalog # 8.22145.0250
Supplier:  Merck
CAS Number:  
Sodium sulphanilate dihydrate sodium salt for synthesis, Sigma-Aldrich®
Catalog # 8.22145.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    170 g/l (20 °C)
  • Molar Mass:
    231.2 g/mol
  • Chemical formula:
    4-(NH2)C6H4SO3Na*2H2O
  • Molecular weight:
    231,2 g/mol
  • Synonyms:
    Sulfanilic acid sodium salt
  • Identity (IR):
    passes test
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
 :231.198 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:208-208-5
CAS Number:6106-22-5
 :MFCD00151042
Formula:C₆H₁₀NNaO₅S
MDL Number:MFCD00064394