Order Entry
Germany
ContactUsLinkComponent
Anti-TAAR8 Rabbit Polyclonal Antibody
Anti-TAAR8 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5060R
Supplier:  Bioss
Anti-TAAR8 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5060R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Trace amine associated receptor 8
  • Antigen symbol:
    TAAR8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    83551
  • Antigen synonyms:
    G-protein coupled receptor 102|GPR102|TRAR5.|Trace amine receptor 5|TAR5|TA5|TaR-5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

FADS1 is a component of a lipid metabolic pathway that catalyzes biosynthesis of highly unsaturated fatty acids from precursor essential polyunsaturated fatty acids, linoleic acid and alphanlinolenic acid. It catalyzes the desaturation of dihomo gamma linoleic acid and eicosatetraenoic acid to generate arachidonic acid and eicosapentaenoic acid respectively.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat