Order Entry
Germany
ContactUsLinkComponent
Anti-ADCY1 Rabbit Polyclonal Antibody
Anti-ADCY1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3681R
Supplier:  Bioss
undefined
Anti-ADCY1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3681R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate Cyclase 1
  • Antigen symbol:
    ADCY1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    107
  • Antigen synonyms:
    AC1|ADCY1|ATP pyrophosphate-lyase 1|DFNB44|Adenylate cyclase type I|Adenylyl cyclase 1|Ca(2+)/calmodulin-activated adenylyl cyclase|Adenylate cyclase type 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    795-845/1118
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

This is a calmodulin-sensitive adenylyl cyclase. May be involved in regulatory processes in the central nervous system. It may play a role in memory acquisition and learning. Plays a role in the regulation of the circadian rhythm of daytime contrast sensitivity probably by modulating the rhythmic synthesis of cyclic AMP in the retina (By similarity).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ADCY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat