Order Entry
Germany
ContactUsLinkComponent
Anti-NUMA Rabbit Polyclonal Antibody
Anti-NUMA Rabbit Polyclonal Antibody
Catalog # BOSSBS-3512R
Supplier:  Bioss
Anti-NUMA Rabbit Polyclonal Antibody
Catalog # BOSSBS-3512R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Mitotic Apparatus Protein 1
  • Antigen symbol:
    NUMA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Gene ID:
    4926
  • Antigen synonyms:
    Nuclear Matrix Protein-22|NMP-22|NUMA1|Centrophilin Stabilizes Mitotic Spindle In Mitotic Cells|NuMA Protein
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

NuMA (Nuclear Mitotic Apparatus Protein) is an intranuclear protein and present in nucleus during interphase. At the onset of mitosis, it redistributes from the nucleus to two centrosomal structures that later will become part of the mitotic spindle pole. After anaphase, the protein redistributes from the spindle polar region into reforming nucleus. NuMA is an essential protein during mitosis for the terminal phases of chromosome separation and/or nuclear reassembly. Recently a study shows that NuMA is cleaved to a 180 to 200kDa during apoptosis.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NUMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat