Anti-HCST Rabbit Polyclonal Antibody
Catalog # BOSSBS-15430R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen symbol:HCST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:PIK3AP|DAP10|Hematopoietic cell signal transducer|phosphoinositide 3 kinase adaptor protein|Membrane protein DAP10|DNAX activation protein 10|Transmembrane adapter protein KAP10|HCST_HUMAN.|KAP10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HCST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human