Anti-Glucoamylase Rabbit Polyclonal Antibody
Catalog # BOSSBS-13383R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Glucoamylase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Form:liquid
- Antigen synonyms:Glucan 1,4|AMYG_ASPAW|Glucoamylase 3|Glucan 14 alpha glucosidase|Amyloglucosidase|Glucoamylase|Glucan 1 4 alpha glucosidase|Gluc 2|1,4-alpha-D-glucan glucohydrolase|1 4 alpha D glucan glucohydrolase|Gluc 1|Gluc 3|Glucoamylase 2|Glucoamylase 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Glucoamylase is an important industrial enzyme used in saccharification steps in both starch enzymatic conversion and in alcohol production. The catalytic domain degrades oligosaccharides from the non reducing end, releasing glucose, and the starch domain binds the enzyme to raw starch and to the cell wall.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Glucoamylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species