Specifications
- Antibody type:Primary
- Antigen name:NOTCH-regulated ankyrin Repeat Protein
- Antigen symbol:NRARP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:NOTCH regulated ankyrin repeat protein|MGC61598|Notch regulated ankyrin repeat containing protein|Notch-regulated ankyrin repeat-containing protein|NRARP_HUMAN.|nrarp
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Nrarp (NOTCH-regulated ankyrin repeat protein) is a 114 amino acid protein that contains two ANK repeats and is thought to play a role in the formation of somites. The gene encoding Nrarp maps to human chromosome 9, which contains 145 million base pairs and comprises 4% of the human genome, encoding nearly 900 genes. Hereditary hemorrhagic telangiectasia and Familial dysautonomia are both associated with chromosome 9. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in translocations that lead to the aberrant production of a BCR-ABL fusion protein often found in leukemias.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: NRARP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat