Order Entry
Germany
ContactUsLinkComponent
Anti-NDUFS7 Rabbit Polyclonal Antibody
Anti-NDUFS7 Rabbit Polyclonal Antibody
  BOSSBS-11734R
 :  Bioss
Anti-NDUFS7 Rabbit Polyclonal Antibody
  BOSSBS-11734R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase)
  • Antigen symbol:
    NDUFS7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    NADH-coenzyme Q reductase|FLJ45860|20kDa (NADH-coenzyme Q reductase)|mitochondrial respiratory chain|FLJ46880|MGC120002|NADH dehydrogenase (ubiquinone) Fe-S protein 7|complex I-20kD|MY017|mitochondrial|NDUFS7|PSST|CI-20KD|complex I|20-KD subunit|NADH:ubiquinone oxidoreductase PSST subunit|NADH-ubiquinone oxidoreductase Fe-S protein 7|NADH dehydrogenase [ubiquinone] iron-sulfur protein 7
  • Storage buffer:
    Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at –20 °C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Located in the mitochondrial inner membrane, mitochondrial complex I is the first and largest enzyme in the electron transport chain of oxidative phosphorylation. By oxidizing NADH that is produced in the Krebs cycle, this complex utilizes the two electrons to reduce ubiquinone to ubiquinol, thereby initiating the passage of electrons to successive complexes and ultimately leading to the reduction of oxygen to water. Mitochondrial complex I consists of over 40 subunits and is of considerable clinical interest since defects in any of the subunits can lead to various myopathies and neuropathies. As a subunit of mitochondrial complex I, NDUFS7 (NADH dehydrogenase [ubiquinone] iron-sulfur protein 7), also designated NADH-ubiquinone oxidoreductase 20 kDa subunit, is a 213 amino acid protein that is suggested to be required for catalytic activity. Defects in the gene encoding NDUFS7 are the cause of Leigh syndrome, a severe neurological disorder that is characterized by bilaterally symmetrical necrotic lesions in subcortical brain regions.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NDUFS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat