Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C motif) Ligand 1, lymphotactin
- Antigen symbol:XCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:XCL1 C mot chemokine 1|ATAC|Lymphotaxin|SCM-1-alpha|Small inducible cytokine C1|Lptn|Small-inducible cytokine C1|Cytokine SCM-1|Xcl1|Lymphotactin|SCM1|SCYC1|Small Inducible Cytokine Subfamily C Member 1|LTN|XCL1_HUMAN.|LPTN|SCM 1 alpha|XC chemokine ligand 1|Chemokine C Mot Ligand 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Lymphotactin is a chemokine-like molecule produced selectively, if not exclusively, by activated CD8+ T cells (1). Lymphotactin is a C-type member of the chemokine superfamily, but retains only the 2nd and 4th of the four cysteine residues conserved in other chemokines (3,4). In normal tissues, lymphotactin is expressed at high levels in spleen, thymus, small intestine, and peripheral blood leukocytes, as well as at low levels in lung, prostate, and ovary (2). Lymphotactin is chemotactic for CD4+ and CD8+ T cells but not for monocytes, and induces a rise in intracellular calcium in peripheral blood lymphocytes (1). The specific receptor for lymphotactin is XCR1, a member of the G protein-coupled receptor family (5). The gene which encodes lymphotactin maps to human chromosome 1q23 (1,2).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: XCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human