Specifications
- Antibody type:Primary
- Antigen name:SLC4A4 variant A
- Antigen symbol:Solute carrier family 4, member 4 variant A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:sodium bicarbonate cotransporter NBCe1 variant A|electrogenic sodium bicarbonate cotransporter 1 isoform 2|SLC4A4 protein.|NBCe1-A|KNBC|hhNMC|NBC2|SLC4A5|HNBC1|pNBC|NBC1|Slc4a4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
SLC4A4 (Electrogenic sodium bicarbonate cotransporter 1) is an electrogenic sodium/bicarbonate cotransporter with a Na(+):HCO3(-) stoichiometry varying from 1:2 to 1:3. It may regulate bicarbonate influx/efflux at the basolateral membrane of cells and regulate intracellular pH. SLC4A4 interacts with carbonic anhydrase 2 and carbonic anhydrase 4 which may regulate transporter activity. There are four named isoforms produced by alternative splicing.This gene encodes a sodium bicarbonate cotransporter (NBC) involved in the regulation of bicarbonate secretion and absorption and intracellular pH. Mutations in this gene are associated with proximal renal tubular acidosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Oct 2008].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: SLC4A4 variant A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat