Anti-Eukaryotic aspartyl protease Rabbit Polyclonal Antibody
BOSSBS-0613R
: Bioss
- Antibody type:Primary
- Antigen name:Eukaryotic aspartyl protease
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Plant
- Form:liquid
- Antigen synonyms:pfam00026|S5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Eukaryotic aspartyl protease
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant