Autopsy blade handles and blades, FEATHER®
Supplier: PFM Medical GmbH
Certificates
About this item
The plastic handle is autoclavable and features excellent chemical resistance. It can be easily disassembled for cleaning. Superior sharpness and cutting performance compared to conventional knives. The same handle can be used for brain dissection and other purposes simply by exchanging the blade.
- Fast and easy blade change
- Handle is easy to clean and autoclavable