Forceps
Straight, blunt, Orange, 180 mm
231-1121
: SEMADENI
POM, orange. These electrically insulating forceps can be universally used.
- Very durable
- Not suitable for use with strong acids and solvents
- Heat resistant up to +130 °C
- Type:Straight, blunt
- Colour:Orange
- Length:180 mm



