Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Reference standards, SV mix, PCB mix and Pesticide mix
Reference standards, SV mix, PCB mix and Pesticide mix
Reference standards, SV mix, PCB mix and Pesticide mix
Lieferant:
Restek
Zertifikate
Expand image
Reference standards, SV mix, PCB mix and Pesticide mix
Lieferant:
Restek
Über diesen Artikel
Spezifikationen
CAS No.:
321-60-8
Formula:
C₁₂H₉F
Description:
2-fluorobiphenyl
Spezifikationen