
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:12245-39-5
:
Formula:C₁₃H₂₀O₂Rh
MDL Number:MFCD00075046
:311.207 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:138 - 140 °C
- Molar Mass:310.2 g/mol
- Assay (to C, H):≥96%
- Identity (IR):Passes test
- Storage temperature:+15...+25 °C