Order Entry
Germany
ContactUsLinkComponent
Methyltrialkylammonium chloride (mixture of C8-C10) for synthesis, Sigma-Aldrich®
Methyltrialkylammonium chloride (mixture of C8-C10) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Methyltrialkylammonium chloride (mixture of C8-C10) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Boiling Point:
    240 °C
  • Flash Point:
    113 °C
  • Density:
    0.89 g/cm3 (20 °C)
  • Assay (argentometric):
    85 - 95%
  • Water (K.F):
    2 - 5%
  • Storage temperature:
    +15...+25 °C
  • Identity (IR):
    Passes test

Specification Test Results

Related Information