ContactUsLinkComponent
Meiosis model
Meiosis model
Catalog # BINH1013869
Supplier:  3B Scientific
undefined
Meiosis model
Catalog # BINH1013869
Supplier:  3B Scientific
Specifications
  • Description:
    Meiosis model
  • Pk:
    1
Specifications
About this item
This meiosis model series shows the ten stages of meiosis on the basis of a typical mammal cell at an enlargement of approximately 10000 times. The three-dimensional relief models are painted according to the usual coloring methods of microscopy, making the process of meiotic cell division easy to understand. The cell organelles are portrayed open in the lower part of the models. The meiosis models are equipped with magnets at the rear so that they can be easily arranged on a magnetic board in the classroom. The detailed stages of meiosis:
  • 1. Interphase, stage of G1-phase
  • 2. Prophase I, leptotene
  • 3. Prophase I, zygotene and pachytene
  • 4. Prophase I, diplotene
  • 5. Prophase I, diakinesis
  • 6. Metaphase I
  • 7. Anaphase I
  • 8. Telophase I, cytokinesis I, interkinesis, prophase II and metaphase II
  • 9. Anaphase II
  • 10. Telophase II and cytokinesis II
Ordering information: Comes complete with a detailed description and handouts to copy for your lessons.
Delivery: Supplied in a storage system (400×600 mm) that can be hung on a wall.