Anti-IFIT1 Rabbit Polyclonal Antibody
ABNOPAB29464
: Abnova
- Antibody type:Primary
- Antigen name:Interferon-induced protein with tetratricopeptide repeats 1
- Antigen symbol:IFIT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Gene ID:3434
- Antigen synonyms:IFI56|RNM561|P56|IFNAI1|IFI-56K|ISG56|C56|IFI-56|IFIT-1|G10P1
- Sequence:KGQPRGQNREKLDKMIRSAIFHFESAVEKKPTFEVAHLDLARMYIEAGNH
- Immunogen:Recombinant protein corresponding to human IFIT1.
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant human IFIT1.
Type: Primary
Antigen: IFIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human