Order Entry
Germany
ContactUsLinkComponent
 
Electronic flow meter, ProFLOW 6000
  REST22656
 :  Restek
undefined
Electronic flow meter, ProFLOW 6000
  REST22656
 :  Restek

 

  • Flow rate:
    0,5 - 500 ml/min
  • Description:
    Flow meter, electronic, ProFLOW 6000, with 0...+48 °C operating temperature
  • Accuracy:
    ±2 %/±0,2 ml/min, whichever greater
  • Pk:
    1

 

 

The ProFLOW 6000 is designed with explosive and flammable gases.

  • Volumetric flow for gases in a range of 0,5 to 500 ml per minute
  • Automatic shut-off function, ergonomic design, side grips
  • Over range warning indicator
 : Supplied with two batteries and a hard sided carrying case.
 : Combustible gases are indispensable due to the increasing use of hydrogen in chromatography.