ContactUsLinkComponent
Solid phase extraction disks, Resprep® Oil & Grease
Solid phase extraction disks, Resprep® Oil & Grease
Catalog # REST26022
Supplier:  Restek
undefined
Solid phase extraction disks, Resprep® Oil & Grease
Catalog # REST26022
Supplier:  Restek
Specifications
  • Ø:
    47 mm
  • Description:
    Solid phase extraction disks, Resprep® Oil & Grease
  • Pk:
    20
Specifications
About this item
Glass fiber disks embedded with specialty bonded silica. These disks reduce the emulsion formation and amount of solvent required by EPA methods. No chlorofluorocarbons needed.
Certifications: Meets the requirements for EPA method 1664.