ContactUsLinkComponent
pH tester, Eutech EcoTestr pH 1 + pH 2
pH tester, Eutech EcoTestr pH 1 + pH 2
Supplier:  EUTECH
Certificates
undefined
pH tester, Eutech EcoTestr pH 1 + pH 2
Supplier:  EUTECH
Specifications
  • Catalog No:
  • 662-2014
  • 662-2015
  • Model:
  • EcoTestr pH 1
  • EcoTestr pH 2
  • pH range:
  • 0,0 - 14,0
  • 0,0 - 14,0
  • pH resolution:
  • 0,1
  • 0,1
  • pH accuracy:
  • ±0,1
  • ±0,1
  • pH calibration:
  • 1 point (automatic)
  • 3 points (automatic)
  • Temperature compensation:
  • -
  • Automatic (ATC)
  • Display:
  • 3½ digit LCD with annunciator
  • 3½ digit LCD with annunciator
  • Power supply:
  • 4 × 1,5 V ‘A76’ micro alkaline batteries
  • 4 × 1,5 V ‘A76’ micro alkaline batteries
  • Compliance:
  • IP67
  • IP67
  • W×D×H:
  • 163×45×30 mm
  • 163×45×30 mm
  • Weight:
  • 0,09 kg
  • 0,09 kg