ContactUsLinkComponent
Inland 45 pump oil
Inland 45 pump oil
Catalog # REST24819
Supplier:  Restek
undefined
Inland 45 pump oil
Catalog # REST24819
Supplier:  Restek
Specifications
  • Description:
    Oil for Inland 45 vacuum pump
  • Pk:
    1 Bottle
Specifications
About this item
Inland 45 pump oil is recommended for most mass spectrometers. It has low vapor pressure,10⁻⁷ torr and optimum vacuum pump performance.
  • Non-toxic and non-corrosive
  • Compatible with buna-N, neoprene, and Viton® seals