Syringe filters, with Luer lock inlet
Supplier: Restek
Certificates
About this item
Syringe filters with hydrophobic PTFE membranes and PP housing are used for organic solvents, acids, alcohols, bases and aromatics.
- Luer lock inlet for leak-tight syringe connection
- Autoclavable at 121 °C for 15 min
- Colour coded for easy identification