Solid phase extraction cartridges for pesticide residue cleanup
Supplier: Restek
Certificates
About this item
Pesticide residue cleanup SPE cartridges are convenient with multiple adsorbent beds in a single cartridge. It is used in multiresidue pesticide analysis to remove matrix interferences and also exhibits good cleanup of dietary supplement extracts.