Orders LinkContactUsLinkComponent
QuEChERS dSPE tubes, Q-sep™
QuEChERS dSPE tubes, Q-sep™
Supplier:   Restek
QuEChERS dSPE tubes, Q-sep™
Supplier:  Restek
Certificates
QuEChERS dSPE tubes, Q-sep™
QuEChERS dSPE tubes, Q-sep™
20 Options Available Below
Product Details & Documents
QuEChERS dSPE tubes are used for extract cleanup. It is fast, simple sample Prep for multiresidue pesticide analysis. It has a advantage that no glassware required and it is ready to use. It can be individually labelled for easy sorbent identification and the tubes are preweighed with ultra pure sorbents.
Support original unbuffered, AOAC (2007.01), european (EN 15662) and mini-multiresidue QuEChERS methods.
Packaging: Packaged in foil subpacks of 10 for enhanced protection and storage stability.
Product Family Options
Micro-centrifuge tube for cleanup of 1 ml extract
Product Information
  • Capacity
    Availability
    Price
  • REST26123
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 235,00 €
    235,00 €
    1 * 100 ST
       235,00 €
  • REST26124
    Tubes with 150 mg magnesium sulphate, 50 mg PSA
    Tubes with 150 mg magnesium sulphate, 50 mg PSA
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • REST26125
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg C18
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg C18
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 150,00 €
    150,00 €
    1 * 100 ST
       150,00 €
  • REST26215
    Tubes with 150 mg magnesium sulphate, 25 mg PSA
    Tubes with 150 mg magnesium sulphate, 25 mg PSA
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 150,00 €
    150,00 €
    1 * 100 ST
       150,00 €
  • REST26216
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 25 mg C18
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 25 mg C18
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 213,00 €
    213,00 €
    1 * 100 ST
       213,00 €
  • REST26217
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 2,5 mg graphitised carbon black
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 2,5 mg graphitised carbon black
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 246,00 €
    246,00 €
    1 * 100 ST
       246,00 €
  • REST26218
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 7,5 mg graphitised carbon black
    Tubes with 150 mg magnesium sulphate, 25 mg PSA, 7,5 mg graphitised carbon black
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 246,00 €
    246,00 €
    1 * 100 ST
       246,00 €
  • REST26219
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black and 50 mg C18
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black and 50 mg C18
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 246,00 €
    246,00 €
    1 * 100 ST
       246,00 €
  • REST26242
    Tubes with 150 mg magnesium sulphate, 50 mg C18
    Tubes with 150 mg magnesium sulphate, 50 mg C18
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 213,00 €
    213,00 €
    1 * 100 ST
       213,00 €
  • REST26243
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 7,5 mg graphitised carbon black and 50 mg C18
    Tubes with 150 mg magnesium sulphate, 50 mg PSA, 7,5 mg graphitised carbon black and 50 mg C18
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 213,00 €
    213,00 €
    1 * 100 ST
       213,00 €
Centrifuge tube for cleanup of 6 ml and 8 ml extract
Product Information
  • Capacity
    Availability
    Price
  • REST26126
    Tubes with 900 mg magnesium sulphate, 300 mg PSA, 150 mg graphitised carbon black
    Tubes with 900 mg magnesium sulphate, 300 mg PSA, 150 mg graphitised carbon black
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 213,00 €
    213,00 €
    1 * 50 ST
       213,00 €
  • REST26220
    Tubes with 1200 mg magnesium sulphate, 400 mg PSA
    Tubes with 1200 mg magnesium sulphate, 400 mg PSA
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26221
    Tube with 1200 mg magnesium sulphate, 400 mg PSA and 400 mg C18
    Tube with 1200 mg magnesium sulphate, 400 mg PSA and 400 mg C18
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26222
    Tube with 1200 mg magnesium sulphate, 400 mg PSA, 400 mg C18 and 400 mg graphitised carbon black
    Tube with 1200 mg magnesium sulphate, 400 mg PSA, 400 mg C18 and 400 mg graphitised carbon black
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 376,00 €
    376,00 €
    1 * 50 ST
       376,00 €
  • REST26223
    Tubes with 900 mg magnesium sulphate, 150 mg PSA
    Tubes with 900 mg magnesium sulphate, 150 mg PSA
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26224
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 15 mg graphitised carbon black
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 15 mg graphitised carbon black
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26225
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 45 mg graphitised carbon black
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 45 mg graphitised carbon black
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26226
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 150 mg C18
    Tubes with 900 mg magnesium sulphate, 150 mg PSA and 150 mg C18
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 143,00 €
    143,00 €
    1 * 50 ST
       143,00 €
  • REST26244
    Tubes with 1200 mg magnesium sulphate, 400 mg C18
    Tubes with 1200 mg magnesium sulphate, 400 mg C18
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 158,00 €
    158,00 €
    1 * 50 ST
       158,00 €
  • REST26245
    Tube with 900 mg magnesium sulphate, 300 mg PSA, 300 mg C18 and 45 mg graphitised carbon black
    Tube with 900 mg magnesium sulphate, 300 mg PSA, 300 mg C18 and 45 mg graphitised carbon black
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 151,00 €
    151,00 €
    1 * 50 ST
       151,00 €