Capillary columns, Rtx®-502.2 (fused silica)
Supplier: Restek
Certificates
About this item
Application-specific columns with unique selectivity for volatile organic pollutants. The Rtx®-502.2 column is cited in U.S. EPA method 502.2 and in many gasoline range organics (GRO) methods for monitoring underground storage tanks. For separation of trihalomethanes, ideal polarity for light hydrocarbons and aromatics.
Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase.
Temperature range: Stable to 270 °C
Specifications
- Catalog No:
- REST10986
- REST10920
- REST10921
- REST10910
- REST10915
- REST40915
- REST10916
- REST40914
- REST10919
- REST10908
- REST10909
- Temperature limit:
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C
- −20...+250/270 °C