Ceramic plates for bunsen burners
Supplier: NEOLAB
Certificates
About this item
These ceramic plates with aluminium frame are the ideal substitution for asbestos bases.
- 5 mm thick plates with skirt
- Temperature resistant up to 950 °C
- Available in two sizes