Capillary columns, Rtx®-5MS (fused silica)
Supplier: Restek
Certificates
About this item
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g. Aroclor mixes), essential oils, semivolatiles. Column specifically tested for low-bleed performance.
Low-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
Temperature range: −60 to 350 ºC
Similar phases: DB-5, HP-5, HP-5MS, Ultra-2, SPB-5, CP-Sil 8 CB, ZB-5.
Equivalent to USP G27 and G36 phases.
Specifications
- Catalog No:
- REST12637
- REST12638
- REST12694-304
- REST12639
- REST12626
- REST12627
- REST12635
- REST12636
- REST12638-101
- REST12621
- REST12623
- REST12624
- REST12609
- REST12611
- REST12620
- REST12605
- REST12624-6850
- REST12655-6850
- REST12606
- REST12626-118
- REST12608
- REST12626-600
- REST12626-6850
- REST12623-600
- REST12624-042
- REST12650-104
- REST12694
- REST12650-123
- REST12695
- REST12690
- REST12691
- REST12623-303
- REST12692
- REST12623-305
- REST12693
- REST12620-600
- REST12641-6850
- REST12623-042
- REST12623-106600
- REST12670
- REST12623-112
- REST12623-124042
- REST12655
- REST12667
- REST12639-106
- REST12652
- REST12653
- REST12654
- REST12639-600
- REST12640
- REST12641
- REST12642
- REST12620-364
- REST12638-337
- REST12605-123
- REST12638-600
- REST12608-600
- Temperature limit:
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C