Capillary columns, Stabilwax®-DB (fused silica)
Supplier: Restek
Certificates
About this item
Application-specific columns for underivatised amines and other basic compounds including alkylamines, diamines, triamines, nitrogen containing heterocyclics.
Polar phase; Crossbond® base-deactivated Carbowax® polyethylene glycol for amines and basic compounds
Temperature range: 40 to 220 °C
Stabilwax®-DB columns reduce adsorption and improve responses for many basic compounds, without analyte derivatisation or column priming.
Specifications
- Catalog No:
- REST10858
- REST10869
- REST10852
- REST10855
- REST10857
- REST10824
- REST10838
- REST10839
- REST10840
- REST10851
- REST10820
- REST10821
- REST10823
- Temperature limit:
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C
- +4,0...+210/220 °C