Injection port liners, Uniliner®
Supplier: Restek
Certificates
About this item
Liners are used for conversion of ¼" packed injection port.
- Versatile, 0,53 mm int.Ø version can be used in the direct (DI) or on-column (OC) injection mode
- Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
- Reduces solvent tailing