Order Entry
Germany
ContactUsLinkComponent
 
Acid green 1 indicator (C.I. 10020), Millipore®
  1.01306.0025
 :  Merck
 :  
Acid green 1 indicator (C.I. 10020), Millipore®
  1.01306.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Solubility:
    160 g/l (20 °C)
  • Molar Mass:
    878.46 g/mol
  • Bulk Density:
    620 kg/m3
  • pH:
    9.3 (10 g/l, H2O, 20 °C)
  • Storage temperature:
    +5...+30 °C

 

 

 :
 :878.45 g/mol
 :M-Clarity™ quality level = MQ200
 :Ambient
 :243-010-2
 :19381-50-1
 :C₃₀H₁₈FeN₃O₁₅S₃
 :MFCD00003886