Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Sorbents
TLC Sorbents
Cellulose microcrystalline for TLC plates
Cellulose microcrystalline for TLC plates
Cellulose microcrystalline for TLC plates
Catalog # 1.02330.0500
Supplier:
Merck
CAS Number:
SDS
Certificates
Expand image
Cellulose microcrystalline for TLC plates
Catalog # 1.02330.0500
Supplier:
Merck
CAS Number:
SDS
Certificates
Specifications
Particle Size:
<20 µm
Pk:
500 g
Description:
Cellulose microcrystalline
Packed:
In plastic
Specifications
About this item
Product Details
Documentation
Product Details
Documentation